Spex LCS-3117-ACN Piperonyl butoxide LC/MS Standard in HPLC Acetonitrile; 1 mL

Representative image only.

Spex LCS-3117-ACN Piperonyl butoxide LC/MS Standard in HPLC Acetonitrile; 1 mL

Spex | Item: 0455852

LC/MS single-component organic standards reduce potential interference and increase analytical accuracy

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm piperonyl butoxide LC/MS organic standard in HPLC acetonitrile matrix
  • 1 mL ampule

More About this Item

Spex CertiPrep® LC/MS single-component standards are created in HPLC and LC/MS grade solvents to reduce potential interference and increase analytical accuracy.

High-performance liquid chromatography (HPLC) is a column-based chromatographic technique used to separate components in a mixture for identification and quantification. A pump system passes pressurized mobile phase and sample through a prefilled column containing a packing or sorbent which aids in the separation of component peaks. As each analyte in the mixture interacts with the column material, it retards the flow of the analytes within the column. HPLC can be used for identification and quantification of compounds in a variety of fields from agriculture to medical and manufacturing.

A minimal HPLC instrument configuration includes a sampler, pumps, column compartment, and a detector. There are a multitude of various detectors commonly in use with a HPLC system including: refractive index detector (RID), UV/Vis detector, photoiode array detector, and mass spectrometers. Each of these detectors have their own range of use and sensitivity, from the broad range RID detector with its higher detection limits, to the powerful and sensitive LC/MS techniques used for extremely low levels of detection.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixHPLC Acetonitrile
  • Element/IonPiperonyl butoxide
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant
  • CAS Number51-03-6
  • UN NumberUN1648
  • Shelf Life (months)36
  • DescriptionPiperonyl butoxide LC/MS Organic Standard, 1,000 µg/mL in HPLC Acetonitrile; 1 mL
Documents & Certificates
Reviews
Q&A