100 ppm dibrom (naled) cannabis standard in LC/MS acetonitrile matrix
1 mL ampule
More About this Item
While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.
Spex CertiPrep® offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.
Designed for state specific pesticide regulations: • OAR 333-008-11 • HB 3460 • AOAC 2007-01 • EN 15662
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Hazardous material shipping fees may apply.
Specifications
Concentration100 µg/mL (100 ppm)
MatrixLC/MS Acetonitrile
Element/IonDibrom (Naled)
FormLiquid
Volume (mL)1
DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
Storage SymbolFreezer
Hazard SymbolFlammable, Irritant
UN NumberUN1648
CAS Number300-76-5
Shelf Life (months)12
DescriptionDibrom (Naled) CannStandard, 100 µg/mL in LC/MS Acetonitrile; 1 mL