Spex KER-BLK Kerosene Blank, 500 mL

Representative image only.

Spex KER-BLK Kerosene Blank, 500 mL

Spex | Item: 0457016

Stable and accurate kerosene blank

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Determine wear metals in engine oils and lubricants
  • ISO 9001 and ISO/IEC 17025 compliant

More About this Item

The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc.

By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • ClassOrganometallic Standards
  • Element/IonKerosene
  • Volume (mL)500
  • Storage SymbolAmbient
  • Hazard SymbolHealth Hazard
  • UN NumberUN1223
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionKerosene Blank, 500 mL
Documents & Certificates
Reviews
Q&A