Spex IS-BUF3-500 Low Level TISAB II Ionic Strength Adjustment Buffer; 500 mL

Representative image only.

Spex IS-BUF3-500 Low Level TISAB II Ionic Strength Adjustment Buffer; 500 mL

Spex | Item: 0456929

Increases the accuracy of the reading with ionic strength adjustment buffers

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Low level total ionic strength adjustment buffer (TISAB) II in H2O matrix
  • 500 mL bottle

More About this Item

Ionic strength adjuster helps reduce measurement error by ensuring that samples and standards have a similar ionic strength.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • MatrixH2O
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquids, flammable, n.o.s (sodium hydroxide solution, acetic acid, glacial); Class 8 and 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Corrosion
  • UN NumberUN2920
  • Shelf Life (months)12
  • DescriptionLow Level TISAB II Ionic Strength Adjustment Buffer in H2O; 500 mL
Documents & Certificates
Reviews
Q&A