Spex INTER5-500 Assurance® Grade Interference Check Standard 5 for AA & ICP in 5% HNO3; 500 mL

Representative image only.

Spex INTER5-500 Assurance® Grade Interference Check Standard 5 for AA & ICP in 5% HNO3; 500 mL

Spex | Item: 0456822

Multi-element standards for AA & ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Set or confirm background correction intervals
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
  • Interference check standard 5 in 5% HNO3 matrix
  • 500 mL bottle

More About this Item

Use these Interference Check Standards to set or confirm that the correct background correction intervals have been set for sequential ICP spectrometers and that the proper inter-element correction factors are set for simultaneous ICP spectrometers.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-element standard contains 5 components.

• Interference check standard 5 is a 500 mL bottle which contains 6,000 µg/mL (6,000 ppm) of Calcium; 5,000 µg/mL (5,000 ppm) of Iron; 3,000 µg/mL (3,000 ppm) of Magnesium; 1,200 µg/mL (1,200 ppm) of Aluminum; and 1,000 µg/mL (1,000 ppm) of Sodium.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Purity GradeAssurance® Grade
  • Matrix5% HNO3
  • Element/IonAluminum, Calcium, Iron, Magnesium, and Sodium
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Interference Check Standard 5 for AA & ICP in 5% HNO3; 500 mL
Documents & Certificates
Reviews
Q&A