Spex INTER18-100 Assurance® Grade Interference Check Standard 18 with Mercury for AA & ICP in 5% HNO3; 125 mL

Representative image only.

Spex INTER18-100 Assurance® Grade Interference Check Standard 18 with Mercury for AA & ICP in 5% HNO3; 125 mL

Spex | Item: 0456823

Multi-element standards for AA & ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Set or confirm background correction intervals
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
  • Interference check standard 18 with mercury in 5% HNO3 matrix
  • 125 mL bottle

More About this Item

Use these Interference Check Standards to set or confirm that the correct background correction intervals have been set for sequential ICP spectrometers and that the proper inter-element correction factors are set for simultaneous ICP spectrometers.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-element standard contains 17 components.

• Interference check standard 18 with mercury is a 125 mL bottle which contains 20,000 µg/mL (20,000 ppm) of Potassium; 1,000 µg/mL (1,000 ppm) of Arsenic, Lead, Thallium; 500 µg/mL (500 ppm) of Selenium; 300 µg/mL (300 ppm) of Barium, Cadmium, Chromium, Cobalt, Copper, Nickel, Silver, Vanadium, Zinc; 200 µg/mL (200 ppm) of Manganese; and 100 µg/mL (100 ppm) of Beryllium, Mercury*.

Hazardous material shipping fees may apply.

*Mercury is supplied as a separate solution in 5% HNO3 due to incompatibility with other elements.

Specifications
  • ConcentrationMultiple
  • Purity GradeAssurance® Grade
  • Matrix5% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion, Health Hazard
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Interference Check Standard 18 with Mercury for AA & ICP in 5% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A