Spex INTER-SETN Assurance® Grade Interference Check Std Set w/o Mercury; 125 mL

Representative image only.

Spex INTER-SETN Assurance® Grade Interference Check Std Set w/o Mercury; 125 mL

Spex | Item: 0456828

Multi-element standards for AA and ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2018
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
  • Interference check standard without mercury for AA & ICP contains 3 solutions
  • Three 125 mL bottles

More About this Item

The Interference Check Standards are used to set or confirm that the correct background correction intervals have been set for sequential ICP spectrometers and that the proper inter-element correction factors are set for simultaneous ICP spectrometers.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard set contains 3 solutions.

• Solution 1 is a 125 mL bottle which contains 20,000 µg/mL (20,000 ppm) of Potassium; 1,000 µg/mL (1,000 ppm) of Arsenic, Lead, Thallium; 500 µg/mL (500 ppm) of Barium, Cadmium, Chromium, Cobalt, Copper, Nickel, Silver, Vanadium, Zinc; 200 µg/mL (200 ppm) of Manganese; and 100 µg/mL (100 ppm) of Beryllium in 5% HNO3. This is available individually as part number 04568-23.

• Solution 2 is a 125 mL bottle which contains 6,000 µg/mL (6,000 ppm) of Calcium; 5,000 µg/mL 5,000 ppm) of Iron; 3,000 µg/mL (3,000 ppm) of Magnesium; 1,200 µg/mL (1,200 ppm) of Aluminum; and 1,000 µg/mL (1,000 ppm) of Sodium in 5% HNO3. This is available individually as part number 04568-21.

• Solution 3 is a 125 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Antimony in H2O/0.6% Tartaric Acid/tr. HNO3. This is available individually as part number 04503-03.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Purity GradeAssurance® Grade
  • MatrixMultiple
  • Element/IonThree bottle set
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion, Health Hazard
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Interference Check Standard Set without Mercury for AA & ICP, 3 Bottle Set; 125 mL
Reviews
Q&A