Spex INT-B2 ICP Alternate Interferents B Standard; 500 mL

Representative image only.

Spex INT-B2 ICP Alternate Interferents B Standard; 500 mL

Spex | Item: 0456922

Multi-element standards for ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Multi-element alternate interferents B standard for ICP in 5% HNO3/tr. Tartaric Acid/tr. HF matrix
  • 500 mL bottle

More About this Item

Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentration levels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:
• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

Alternate interferents A (04569-21) and alternate analytes B (04569-22) may be prepared in combination with the Alternate interferents A (04569-21) and Analytes B3 (04569-08) solutions mentioned, or any combination involving the four solutions, depending on the analytes and interferents of interest to you. We provides ICP-MS interferents and interferent check solutions for SW-845.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 12 elements.

• ICP Alternate Interferents B standard is a 500 mL bottle which contains 100 µg/mL (100 ppm) Aluminum, Antimony, Arsenic, Boron, Molybdenum, Selenium, Sodium, Thallium and 10 µg/mL (10 ppm) Calcium, Iron, Magnesium, Silicon.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionICP Alternate Interferents B Standard in 5% HNO3/tr. Tartaric Acid/tr. HF; 500 mL
Documents & Certificates
Reviews
Q&A