Spex INT-B1 ICP Analytes B1 Standard in 5% HNO3; 125 mL

Representative image only.

Spex INT-B1 ICP Analytes B1 Standard in 5% HNO3; 125 mL

Spex | Item: 0456909

CLP ILM 02.0 standard for ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Use with 04569-07 to prepare the most common interference check solution
  • 125 mL bottle

More About this Item

Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by SPEX CertiPrep for convenience of use and stability.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentrationlevels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:

• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

Use this standard for verification of inter-element and background correction factors at the beginning and the end of each analysis run. In addition, a verification must be done after every 20th sample. Two solutions are required for the most common interference check: Solution A, the interferents alone (04569-07) and Solution AB, a combination of interferents (04569-07) and analytes (04569-09). Solution A is prepared by diluting ICP Interferents A1 (04569-07) 20-fold. Solution AB is prepared by diluting ICP Interferents A1 (04569-07) 20-fold and ICP Analytes B1 (04569-09) 100-fold; for example, 5 mL of ICP Interferents A1 (04569-07) and 1 mL of ICP Interferents A1 (04569-07) into a 100 mL volumetric flask, brought to volume with a matrix blank. Once prepared, the solutions should be analyzed consecutively, starting with Solution A.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 12 elements.

• Analytes B1 standard is a 125 mL bottle of 100 µg/mL (100 ppm) Cadmium, Lead, Nickel, Silver, Zinc and 50 µg/mL (50 ppm) Barium, Beryllium, Chromium, Cobalt, Copper, Manganese, Vanadium in 5% HNO3.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionICP Analytes B1 Standard in 5% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A