Spex Infinitas™ IFC-K-1A-250 Potassium Nitrate, = 99.99% Trace Metal Basis, 250 g

Representative image only.

Spex Infinitas™ IFC-K-1A-250 Potassium Nitrate, = 99.99% Trace Metal Basis, 250 g

Spex | Item: 5013348

When purity is critical and the slightest impurity matters, trust Infinitas™ to deliver the highest purity chemicals for your most demanding applications

Call For Price

Call For Price
Product Description

Key Features

  • Unrivaled Precision: Perfect for sensitive chemical synthesis and catalysis, where even the smallest impurity can disrupt critical reactions
  • Cutting-Edge Manufacturing: Ideal for high-tech industries like electronics, semiconductors, and batteries, where purity ensures peak performance and reliability
  • Regulatory Excellence: Ensure compliance and quality in food, pharmaceutical, and supplement production by using chemicals that meet the most stringent purity standards

More About this Item

Certain industries demand the absolute highest level of chemical purity. Whether it's food, pharmaceuticals, electronics, or semiconductors, even trace impurities can have significant consequences, from health risks to product failures.

Infinitas™ is your solution. With over 75 years of expertise in providing high-purity standards for the analytical laboratory industry, we now offer fine inorganic chemicals with purity levels exceeding 99.99%. Our products are meticulously tested to ensure impurities are measured down to the ppb and ppm levels.

Specifications
  • Element/IonPotassium nitrate
  • FormCrystalline
  • Weight (g)250
  • CAS Number7757-79-1
  • DescriptionPotassium Nitrate, ≥ 99.99% Trace Metal Basis, 250 g
Documents & Certificates
Reviews
Q&A