Spex ICP-KIT-1 Assurance® Grade Standards Kit, 1,000 µg/mL (1,000 ppm) for Inductively Coupled Plasma (ICP) Analysis, 125 mL

Representative image only.

Spex ICP-KIT-1 Assurance® Grade Standards Kit, 1,000 µg/mL (1,000 ppm) for Inductively Coupled Plasma (ICP) Analysis, 125 mL

Spex | Item: 0456752

Set of 38 single-element standards

Call For Price

Call For Price
Product Description

Includes

38 single-element standards.

Key Features

  • 1,000 µg/mL standards
  • Meant for use in ICP analysis
  • Standards for inductively coupled plasma analysis

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Custom standards can be manufactured upon request.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Irritant
Specifications
  • Concentration1,000 µg/mL (1,000 ppm) for each element
  • Purity GradeAssurance® Grade
  • MatrixSilver, Aluminum, Arsenic, Barium, Beryllium, Calcium, Cadmium, Cobalt, Chromium, Copper, Iron, Potassium, Lithium, Magnesium, Manganese, Sodium, Nickel, Lead, Scandium, Selenium, Strontium, Thallium, Vanadium, Yttrium, Zinc, Zirconium in 2% HNO3 - Bismuth, Mercury in 10% HNO3 - Tin in 20% HCl - Boron, Molybdenum, Phosphorus, Sulfur, Tungsten in H2O - Antimony in H2O, 0.6% tartaric acid, and tr. HNO3 - Titanium in H2O and 0.24% F(-) - Niobium, Silicon in H2O and 0.4% F(-)
  • ClassInorganic Standards
  • Element/IonSilver, Aluminum, Arsenic, Barium, Beryllium, Calcium, Cadmium, Cobalt, Chromium, Copper, Iron, Potassium, Lithium, Magnesium, Manganese, Sodium, Nickel, Lead, Scandium, Selenium, Strontium, Thallium, Vanadium, Yttrium, Zinc, Zirconium, Bismuth, Mercury, Tin, Boron, Molybdenum, Phosphorus, Sulfur, Tungsten, Antimony, Titanium, Niobium, Silicon
  • Volume (mL)125
  • DOT InformationHazmat Fees May Apply
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionStandards Kit, 1,000 µg/mL (1,000 ppm) for Inductively Coupled Plasma (ICP) Analysis, 125 mL
Documents & Certificates
Reviews
Q&A