Spex ICMIX6-100 IC Instrument Ion Anion Check Standard 6 in H2O; 125 mL

Representative image only.

Spex ICMIX6-100 IC Instrument Ion Anion Check Standard 6 in H2O; 125 mL

Spex | Item: 0456932

Multi-ion standards for ion chromatography

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Traceable standards
  • Perfect for the analytical laboratory

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. To ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep®offers the highest quality IC standards available for the analytical laboratory.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • MatrixH2O
  • Element/IonSulfate, Bromide, Chloride, Nitrate, Nitrite, Fluoride
  • FormLiquid
  • Volume (mL)125
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • UN NumberNot regulated
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionIC Instrument Ion Anion Check Standard 6 in H2O; 125 mL
Documents & Certificates
Reviews
Q&A