Spex ICMIX1-100 IC Instrument Ion Anion Check Standard 1 in H2O; 125 mL

Representative image only.

Spex ICMIX1-100 IC Instrument Ion Anion Check Standard 1 in H2O; 125 mL

Spex | Item: 0456930

Multi-element anion standard for ion chromatography

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Ideal for calibrating and performing QA/QC checks on ion chromatography systems
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Ion chromatography anion check standard 1 in H2O matrix
  • 125 mL bottle

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep®IC standards are of the highest quality available for the analytical laboratory.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 5 components.

• IC instrument anion check standard 1 is a 125 mL bottle which contains 150 µg/mL (150 ppm) of hydrogen phosphate and sulfate, 100 µg/mL (100 ppm) of nitrate, 30 µg/mL (30 ppm) of chloride, and 20 µg/mL (20 ppm) of fluoride.

Specifications
  • ConcentrationMultiple
  • MatrixH2O
  • Element/IonChloride, Fluoride, Hydrogen phosphate, Nitrate, and Sulfate
  • FormLiquid
  • Volume (mL)125
  • DOT InformationNon-Hazardous; Not regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionIC Instrument Ion Anion Check Standard 1 in H2O; 125 mL
Documents & Certificates
Reviews
Q&A