Spex IC-ELCON2-100 Sodium Bicarbonate (NaHCO3) Eluent Concentrate, 0.5 M in H2O; 125 mL

Representative image only.

Spex IC-ELCON2-100 Sodium Bicarbonate (NaHCO3) Eluent Concentrate, 0.5 M in H2O; 125 mL

Spex | Item: 0456937

Eluent concentrate for ion chromatography

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Made from high-purity salts and filtered ASTM Type I Water
  • 0.5 M sodium bicarbonate eluent concentrate in H2O matrix
  • 125 mL bottle

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep®IC standards are of the highest quality available for the analytical laboratory.

All eluents are at 100-fold concentration and ready for dilution, as needed, with filtered ASTM Type I Water.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • Concentration0.5 M
  • MatrixH2O
  • Element/IonSodium Bicarbonate (NaHCO3)
  • FormLiquid
  • Volume (mL)125
  • DOT InformationNon-Hazardous; Not regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionSodium Bicarbonate (NaHCO3) Eluent Concentrate, 0.5 M in H2O; 125 mL
Documents & Certificates
Reviews
Q&A