Spex GAS-20 Regular Unleaded Gas (87 Octane), 20 mg/mL in Methanol-P&T; 1 mL

Representative image only.

Spex GAS-20 Regular Unleaded Gas (87 Octane), 20 mg/mL in Methanol-P&T; 1 mL

Spex | Item: 0456640

Volatile organic standard for the monitoring of water, soil, and other environmental matrices

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 20,000 ppm regular unleaded gas in methanol-P&T matrix

More About this Item

Use unleaded gas standard for monitoring gasoline intrusion or pollution in the environment including water, soil, and other environmental matrices.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration20 mg/mL (20,000 ppm)
  • MatrixMethanol-P&T
  • Element/IonRegular Unleaded Gas-87 Octane
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion, Irritant, Health Hazard
  • CAS Number86290-81-5
  • UN NumberUN1230
  • Shelf Life (months)36
  • DescriptionRegular Unleaded Gas (87 Octane), 20 mg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A