Spex EPA-SETN Assurance® Grade Environmental EPA Set without Mercury; 125 mL

Representative image only.

Spex EPA-SETN Assurance® Grade Environmental EPA Set without Mercury; 125 mL

Spex | Item: 0456820

Multi-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2017
  • For use in US EPA Method 6010 and 200.7 (Revision 4.4)
  • Environmental EPA set without mercury for AA & ICP contains 10 solutions
  • Eight 125 mL and two 500 mL bottles

More About this Item

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard set contains 10 solutions.

• Solution 1 is a 125 mL bottle which contains 500 µg/mL (500 ppm) of Lead; 200 µg/mL (200 ppm) of Selenium; 150 µg/mL (150 ppm) of Cadmium, Zinc; 100 µg/mL of Manganese; and 50 µg/mL (50 ppm) of Beryllium in 2% HNO3. This is available individually as part number 04567-76.

• Solution 2 is a 125 mL bottle which contains 10,000 µg/mL (10,000 ppm) of Iron and 100 µg/mL (100 ppm) of Barium, Cobalt, Copper, Vanadium in 5% HNO3. This is available individually as part number 04567-78.

• Solution 3 is a 125 mL bottle which contains 500 µg/mL (500 ppm) of Arsenic and 100 µg/mL (100 ppm) of Molybdenum, Silicon in 2% HNO3/tr. HF. This is available individually as part number 04567-80.

• Solution 4 is a 125 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Aluminum; 400 µg/mL (400 ppm) of Potassium; 200 µg/mL (200 ppm) of Aluminum, Sodium; and 20 µg/mL (20 ppm) of Chromium, Nickel in 5% HNO3. This is available individually as part number 04567-82.

• Solution 5 is a 125 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Magnesium; 200 µg/mL (200 ppm) of Antimony, Thallium; 100 µg/mL (100 ppm) of Boron; and 50 µg/mL (50 ppm) of Silver in 5% HNO3/tr. Tartaric Acid/tr. HF. This is available individually as part number 04567-84.

• Solution 6 is a 125 mL bottle which contains 20,000 µg/mL (20,000 ppm) of Potassium; 1,000 µg/mL (1,000 ppm) of Arsenic, Lead, Thallium; 500 µg/mL (500 ppm) of Barium, Cadmium, Chromium, Cobalt, Copper, Nickel, Silver, Vanadium, Zinc; 200 µg/mL (200 ppm) of Manganese; and 100 µg/mL (100 ppm) of Beryllium in 5% HNO3. This is available individually as part number 04568-23.

• Solution 7 is a 125 mL bottle which contains 6,000 µg/mL (6,000 ppm) of Calcium; 5,000 µg/mL 5,000 ppm) of Iron; 3,000 µg/mL (3,000 ppm) of Magnesium; 1,200 µg/mL (1,200 ppm) of Aluminum; and 1,000 µg/mL (1,000 ppm) of Sodium in 5% HNO3. This is available individually as part number 04568-21.

• Solution 8 is a 125 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Antimony in H2O/0.6% Tartaric Acid/tr. HNO3. This is available individually as part number 04503-03.

• Solution 9 is a 500 mL bottle of Hydrochloric acid blank in 5% HCl. This is available individually as part number 04567-47.

• Solution 10 is a 500 mL bottle of Nitric acid blank in 5% HNO3. This is available individually as part number 04567-46.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Purity GradeAssurance® Grade
  • MatrixMultiple
  • Element/Ion10 bottle set
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Environmental EPA Set without Mercury for AA & ICP, 10 Bottle Set; 125 mL
Documents & Certificates
Reviews
Q&A