Spex EP-8-500N Assurance® Grade Primary Drinking Water Metals w/o Mercury; 500mL

Representative image only.

Spex EP-8-500N Assurance® Grade Primary Drinking Water Metals w/o Mercury; 500mL

Spex | Item: 0456836

Multi-element drinking water pollutant standards for AA & ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Refer to US National Primary Drinking Water Regulations 40 CFR, Part 141
  • Primary drinking water metals standard without mercury for AA & ICP in 2% HNO3 matrix
  • 500 mL bottle

More About this Item

Use these standards with procedures for compliance monitoring of drinking water and for analysis of ground and surface water where determination of the drinking water contamination levels are required.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 7 elements.

• Primary Drinking Water Metals Standard is a 500 mL bottle which contains 100 µg/mL (100 ppm) of Barium; 10 µg/mL (10 ppm) of Arsenic, Chromium, Lead, Silver; and 5 µg/mL (5 ppm) of Cadmium, Selenium.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Purity GradeAssurance® Grade
  • Matrix2% HNO3
  • Element/IonArsenic, Barium, Cadmium, Chromium, Lead, Selenium, and Silver
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Grup III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionSpex CertiPrep Assurance® Grade Primary Drinking Water Metals without Mercury for AA & ICP in 2% HNO3; 500 mL
Documents & Certificates
Reviews
Q&A