Spex ENC-EVEN-1K-PK C16 through C36 Even Alkanes Mix, 1,000 µg/mL; 1 mL; 4/pk

Representative image only.

Spex ENC-EVEN-1K-PK C16 through C36 Even Alkanes Mix, 1,000 µg/mL; 1 mL; 4/pk

Spex | Item: 0500955

Test for groundwater and soil contamination caused by underground storage tanks

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm C16 through c36 even alkanes mix in methylene chloride matrix
  • Test groundwater and soil for petroleum or other hazardous substance leakage by underground storage tanks
  • Four 1 mL ampules

More About this Item

The EPA regulates the monitoring and testing of underground storage tanks (USTs), that were or are used to store petroleum or other hazardous substances, which could impact the environment and human health. Currently, there are over half a million registered underground storage tanks in the US alone. Many tanks installed prior to the mid-80s were composed of steel, which can corrode over time allowing for leakage into the surrounding groundwater and soil.

The Hazardous and Solid Waste Amendments to the Resource Conservation and Recovery Act (RCRA), passed in 1984, required the US Environmental Protection Agency (EPA) to develop regulations for the underground storage of motor fuels. The EPA requires owners and operators of USTs to maintain and clean up sites damaged by petroleum contamination.

Spex CertiPrep® provides standards designed to meet the testing requirements set up by the EPA and many individual state regulations for leaking underground storage tanks (LUST) and leaking underground fuel tanks (LUFT).

This multi-component standard contains 11 components.

• C16 through C36 Even Alkanes Mix is a 1 mL ampule which contains 1,000 µg/mL (1,000 ppm) of n-Docosane, n-Dotriacontane, n-Eicosane, n-Hexacosane, n-Hexadecane, n-Hexatriacontane, n-Octacosane, n-Octadecane, n-Tetracosane, n-Tetratriacontane, and n-Triacontane.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryOrganics;Standards
  • Element/IonPhenol-d6
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number13127-88-3
  • UN NumberUN1230
  • Shelf Life (months)36
  • DescriptionSpex CertiPrep Phenol-d6 Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A