Spex ECS-Z-002 Base/Neutral Surrogate Mix (3 Components), 1,000 µg/mL; 11.5 mL

Representative image only.

Spex ECS-Z-002 Base/Neutral Surrogate Mix (3 Components), 1,000 µg/mL; 11.5 mL

Spex | Item: 0456705

Analytical standard for GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm base/neutral surrogate mix with 3 components in methylene chloride matrix
  • For US EPA Methods 625 and 8270
  • 11.5 mL bottle

More About this Item

All ampulized standards are supplied with a prelabeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date. Custom mixes are also available.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 3 components.

• Base/Neutral Surrogate Mix is a 11.5 mL bottle which contains 1,000 µg/mL (1,000 ppm) of 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethylene Chloride
  • Element/Ion2-Fluorobiphenyl, Nitrobenzene-d5, p-Terphenyl-d14
  • FormLiquid
  • Volume (mL)11.5
  • DOT InformationHazardous; Dichloromethane; Class 6.1, Packing Group III
  • Storage SymbolRefrigerator
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1593
  • CAS Number321-60-8, 4165-60-0, 1718-51-0
  • Shelf Life (months)49
  • DescriptionBase/Neutral Surrogate Mix with 3 components, 1,000 µg/mL in Methylene Chloride; 11.5 mL
Documents & Certificates
Reviews
Q&A