Spex ECS-N-008 Decafluorotriphenylphosphine (DFTPP) for US EPA Methods 625 and 8270, 50 µg/mL; 5.5 mL

Representative image only.

Spex ECS-N-008 Decafluorotriphenylphosphine (DFTPP) for US EPA Methods 625 and 8270, 50 µg/mL; 5.5 mL

Spex | Item: 0456722

Analytical standards for GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 50 ppm decafluorotriphenylphosphine (DFTPP) in methylene chloride matrix
  • For US EPA Method 625 and 8270
  • 5.5 mL bottle

More About this Item

All ampulized standards are supplied with a prelabeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration50 µg/mL (50 ppm)
  • MatrixMethylene Chloride
  • Element/IonDecafluorotriphenylphosphine (DFTPP)
  • FormLiquid
  • Volume (mL)5.5
  • DOT InformationHazardous; Dichloromethane; Class 6.1, Packing Group III
  • Storage SymbolRefrigerator
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • CAS Number5074-71-5
  • UN NumberUN1593
  • Shelf Life (months)36
  • DescriptionDecafluorotriphenylphosphine (DFTPP) for US EPA Methods 625 and 8270, 50 µg/mL in Methylene Chloride; 5.5 mL
Documents & Certificates
Reviews
Q&A