Spex ECS-K-050 Semivolatiles Calibration Kit, 2,000 µg/mL, 1.8 mL; 5/PK

Representative image only.

Spex ECS-K-050 Semivolatiles Calibration Kit, 2,000 µg/mL, 1.8 mL; 5/PK

Spex | Item: 6102191

Analytical standard for GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 2,000 ppm semivolatiles calibration kit
  • For US EPA Methods 625 and 8270
  • Kit contains five 1.8 mL vials

More About this Item

All ampulized standards are supplied with a prelabeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date. Custom mixes are also available.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-compound kit contains 5 standards.

• Base/Neutrals Mix 1 is a 1.8 mL vial which contains 2,000 µg/mL (2,000 ppm) of 1,2,4-Trichlorobenzene, 1,2-Dichlorobenzene, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, 2,6-Dinitrotoluene, 2-Chloronaphthalene, 4-Bromodiphenyl ether, 4-Chlorophenyl-phenyl ether, Azobenzene, Bis(2-chloro-1-methylethyl) ether, bis(2-Chloroethoxy)methane, bis(2-Chloroethyl)ether, Bis(2-Ethylhexyl)phthalate, Butylbenzyl phthalate, Carbazole, Diethyl phthalate, Dimethyl phthalate, Di-n-butyl phthalate, Di-n-octyl phthalate, Hexachlorobenzene, Hexachlorobutadiene, Hexachlorocyclopentadiene, Hexachloroethane, Isophorone, Nitrobenzene, N-Nitrosodimethylamine, N-Nitrosodi-n-propylamine, N-Nitrosodiphenylamine, and Pyridine. This is available individually as part number 04567-23.

• Add-Ons Mix 1 for US EPA Methods 625 and 8270 is a 1.8 mL vial which contains 2,000 µg/mL (2,000 ppm) of 2,4,5-Trichlorophenol, 2-Methylnaphthalene, 2-Methylphenol, 2-Nitroaniline, 3-Nitroaniline, 4-Chloroaniline (p-Chloroaniline), 4-Methylphenol, 4-Nitroaniline, Aniline, Benzoic acid, Benzyl alcohol, and Dibenzofuran. This is available individually as part number 04567-35.

• Polynuclear Aromatic Hydrocarbon (PAH) Mix is a 1.8 mL vial which contains 2,000 µg/mL (2,000 ppm) of Acenaphthene, Acenaphthylene, Anthracene, Benz(a)anthracene, Benzo(a)pyrene, Benzo(b)fluoranthene, Benzo(g,h,i)perylene, Benzo(k)fluoranthene, Chrysene, Dibenz(a,h)anthracene, Fluoranthene, Fluorene, Indeno(1,2,3-c,d)pyrene, Naphthalene, Phenanthrene, and Pyrene. This is available individually as part number 04567-32.

• Phenols Mix is a 1.8 mL vial which contains 2,000 µg/mL (2,000 ppm) of 2,4,6-Trichlorophenol, 2,4-Dichlorophenol, 2,4-Dimethylphenol, 2,4-Dinitrophenol, 2-Chlorophenol, 2-Nitrophenol, 4,6-Dinitro-2-methylphenol, 4-Chloro-3-methylphenol, 4-Nitrophenol, Pentachlorophenol, and Phenol. This is available individually as part number 04567-29.

• Benzidines Mix is a 1.8 mL vial wich contains 2,000 µ g/mL (2,000 ppm) of 3,3'-Dichlorobenzidine and Benzidine. This is available individually as part number 04567-26.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMultiple
  • Element/IonVial 1: Base/Neutral Mix 1
    Vial 2: Add-Ons Mix 1
    Vial 3: PAH Mix
    Vial 4: Phenols Mix
    Vial 5: Benzidines Mix
  • FormLiquid
  • Volume (mL)1.8
  • Packaging5 x 1.8 mL
  • DOT InformationHazardous; Dichloromethane; Class 6.1, Packing Group III
  • Storage SymbolRefrigerator
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1593
  • CAS NumberMultiple
  • Shelf Life (months)12
  • DescriptionSemivolatiles Calibration Kit, 2,000 µg/mL, 1.8 mL; 5/PK
Reviews
Q&A