Spex ECS-A-041 ISTD Mix for US EPA Method 8260, 2,000 µg/mL in Methanol-P&T, 1.5 mL

Representative image only.

Spex ECS-A-041 ISTD Mix for US EPA Method 8260, 2,000 µg/mL in Methanol-P&T, 1.5 mL

Spex | Item: 0456676

Analytical standard for GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 2,000 ppm ISTD mix in methanol-P&T matrix
  • For US EPA 8260
  • One 1.5 mL ampule

More About this Item

All ampulized standards are supplied with a prelabeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date. Custom mixes are also available.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 4 components.

• ISTD Mix for US EPA Method 8260 is a 1.5 mL ampule which contains 2,000 µg/mL (2,000 ppm) of Chlorobenzene-d5, 1,4-Dichlorobenzene-d4, 1,4-Difluorobenzene, and Pentafluorobenzene.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • Element/IonChlorobenzene-d5, 1,4-Dichlorobenzene-d4, 1,4-Difluorobenzene, Pentafluorobenzene
  • FormLiquid
  • Volume (mL)1.5
  • Packaging1 x 1.5 mL
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number3114-55-4, 3855-82-1, 540-36-3, 363-72-4
  • UN NumberUN1230
  • Shelf Life (months)36
  • DescriptionISTD Mix for US EPA Method 8260, 2,000 µg/mL in Methanol-P&T, 1.5 mL; 1/EA
Documents & Certificates
Reviews
Q&A