Spex ECS-A-034-PK ISTD Surrogate Mix for EPA Method 524.2, 2,000 µg/mL, 1.5 mL; 4/Pk

Representative image only.

Spex ECS-A-034-PK ISTD Surrogate Mix for EPA Method 524.2, 2,000 µg/mL, 1.5 mL; 4/Pk

Spex | Item: 0500977

ECS Standards for GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 524.2
  • 2,000 ppm ISTD surrogate mix in methanol-P&T matrix
  • Three 1.5 mL ampules

More About this Item

All ampulized standards are supplied with a pre-labeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date. Custom mixes are also available.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryOrganics;Standards
  • Element/IonTetramethylammonium hydroxide
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number75-59-2
  • UN NumberUN1230
  • Shelf Life (months)12
  • DescriptionTetramethylammonium Hydroxide Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A