Spex ECS-A-017-PK 4-Bromofluorobenzene BFB Tune Check Standard, 50 µg/mL, 1.5 mL; 4/Pk

Representative image only.

Spex ECS-A-017-PK 4-Bromofluorobenzene BFB Tune Check Standard, 50 µg/mL, 1.5 mL; 4/Pk

Spex | Item: 0500972

ECS Standards for GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 624, 8240, 8260 and 524.2
  • 50 ppm 4-bromofluorobenzene BFB tune check standard in methanol-P&T
  • One 1.5 mL ampule

More About this Item

All ampulized standards are supplied with a pre-labeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date. Custom mixes are also available.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol
  • ChemistryOrganics;Standards
  • Element/IonBenzidine and 3,3’-Dichlorobenzidine
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Irritant, Health Hazard
  • UN NumberUN1230
  • CAS Number92-87-5, 91-94-1
  • Shelf Life (months)36
  • DescriptionBenzidines Mix for US EPA Methods 605, 8270 & CLP Series Methods, 2,000 µg/mL; 1 mL
Documents & Certificates
Reviews
Q&A