Spex DCBP-XH PCB Screening Standard for US EPA Method 8270, 5,000 µg/mL; 1 mL

Representative image only.

Spex DCBP-XH PCB Screening Standard for US EPA Method 8270, 5,000 µg/mL; 1 mL

Spex | Item: 0455731

Analytical standard for the analysis of semivolatiles in envoronmental and waste samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 5,000 ppm PCB screening standard in toluene matrix
  • 1 mL ampule

More About this Item

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration5,000 µg/mL (5,000 ppm)
  • MatrixToluene
  • Element/IonDecachlorobiphenyl
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Toluene; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1294
  • CAS Number2051-24-3
  • Shelf Life (months)36
  • DescriptionPCB Screening Standard for US EPA Method 8270, 5,000 µg/mL; 1 mL
Documents & Certificates
Reviews
Q&A