Spex CRM-PVCBLK Polyvinyl Chloride (PVC) Phthalate Blank; 1.5 g

Representative image only.

Spex CRM-PVCBLK Polyvinyl Chloride (PVC) Phthalate Blank; 1.5 g

Spex | Item: 0456214

Consumer safety standard helps meet guidelines set by agencies to protect consumers

£51.00GBP / Each
£61.20GBP (incl VAT)

£51.00GBP / Each
£61.20GBP (incl VAT)
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • Use for testing PVC phthalates 04560-99 and 04561-01
  • 1.5 g

More About this Item

Polyvinyl chloride (PVC) is a very common plastic used in a wide range of common consumer products from children’s toys and care items to building and construction materials. In the US, ASTM (American Society for Testing and Materials) and CPSC (Consumer Product Safety Commission) have designed methods for testing children’s toys and childcare articles for compliance with the restriction of use for eight designated phthalates.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • MatrixPolyvinyl Chloride
  • Element/IonPolyvinyl Chloride Blank
  • FormLiquid
  • Weight (g)1.5
  • DOT InformationNot Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot Regulated
  • CAS Number9002-86-2
  • Shelf Life (months)24
  • DescriptionPolyvinyl Chloride (PVC) Phthalate Blank; 1.5 g
Documents & Certificates
Reviews
Q&A