Spex CRM-PVC001 Phthalates Polyvinyl Chloride Standard; 1.5 g

Representative image only.

Spex CRM-PVC001 Phthalates Polyvinyl Chloride Standard; 1.5 g

Spex | Item: 0456099

Consumer safety standard helps meet guidelines set by agencies to protect consumers

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For test method CPSC-CH-C1001-09.1

More About this Item

Polyvinyl chloride, or PVC, is a very common plastic used in a wide range of common consumer products from children’s toys and care items to building and construction materials. In the US, ASTM (American Society for Testing and Materials) and CPSC (Consumer Product Safety Commission) have designed methods for testing children’s toys and childcare articles for compliance with the restriction of use for eight designated phthalates.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 8 components.

• Phthalates Polyvinyl Chloride Standard is a 1.5 g ampule which contains 30,000 µg/g (30,000 ppm) of Diisodecyl phthalate and Diisononyl phthalate; and 3,000 µg/g (3,000 ppm) of Butylbenzyl phthalate, Diethyl phthalate, Dimethyl phthalate, Di-n-butyl phthalate, Di-n-octyl phthalate, and bis(2-Ethylhexyl)phthalate.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixPolyvinyl Chloride
  • Element/IonButylbenzyl phthalate, Diethyl phthalate, Diisodecyl phthalate, Diisononyl phthalate, Dimethyl phthalate, Di-n-butyl phthalate, Di-n-octyl phthalate, bis(2-Ethylhexyl)phthalate
  • FormSolid
  • Weight (g)1.5
  • DOT InformationNot Regulated
  • Storage SymbolAmbient
  • Hazard SymbolHealth Hazard
  • UN NumberNot Regulated
  • CAS Number85-68-7, 84-66-2, 26761-40-0, 28553-12-0, 131-11-3, 84-74-2, 117-84-0, 117-81-7
  • Shelf Life (months)24
  • DescriptionPhthalates Polyvinyl Chloride Standard; 1.5 g
Documents & Certificates
Reviews
Q&A