Spex CRM-PEBLK Polyethylene QC Standard Blank; 5 g

Representative image only.

Spex CRM-PEBLK Polyethylene QC Standard Blank; 5 g

Spex | Item: 0456215

Consumer safety standard helps meet guidelines set by agencies to protect consumers

£58.00GBP / Each
£69.60GBP (incl VAT)

£58.00GBP / Each
£69.60GBP (incl VAT)
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • Blank—to be used for testing PE phthalates

More About this Item

Polyethylene (PE) is one of the world’s most common plastics. PE is used in a variety of common consumer products including children’s toys and care items. Current US regulations limit the concentrations of certain phthalates in childcare articles and children’s toys. Laboratories are tasked with the analysis of children’s toys for these potentially hazardous phthalates.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 8 components.

• Polyethylene QC Standard Blank is a 5 g ampule which contains Bis(2-Ethylhexyl)phthalate, Butylbenzyl phthalate, Diethyl phthalate, Diisodecyl phthalate, Diisononyl phthalate, Dimethyl phthalate, Di-n-butyl phthalate, and Di-n-octyl phthalate.

Specifications
  • Concentration0 (blank)
  • MatrixPolyethylene
  • Element/IonBis(2-Ethylhexyl)phthalate, Butylbenzyl phthalate, Diethyl phthalate, Diisodecyl phthalate, Diisononyl phthalate, Dimethyl phthalate, Di-n-butyl phthalate, Di-n-octyl phthalate
  • FormSolid
  • Weight (g)5
  • DOT InformationNot Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • CAS Number117-81-7, 85-68-7, 84-66-2, 26761-40-0, 28553-12-0, 131-11-3, 84-74-2, 117-84-0
  • UN NumberNot Regulated
  • Shelf Life (months)36
  • DescriptionPolyethylene QC Standard Blank; 5 g
Documents & Certificates
Reviews
Q&A