Spex CRM-PE001 Phthalates Polyethylene Standard in Medium Density Polyethylene; 5 g

Representative image only.

Spex CRM-PE001 Phthalates Polyethylene Standard in Medium Density Polyethylene; 5 g

Spex | Item: 0455998

Consumer safety standard helps meet guidelines set by agencies to protect consumers

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For test method CPSC-CH-C1001-09.1

More About this Item

Polyethylene (PE) is one of the world’s most common plastics. PE is used in a variety of common consumer products including children’s toys and care items. Current US regulations limit the concentrations of certain phthalates in childcare articles and children’s toys. Laboratories are tasked with the analysis of children’s toys for these potentially hazardous phthalates.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 8 components.

• Phthalates Polyethylene Standard is a 5 g ampule which contains 30,000 µg/g (30,000 ppm) of Diisodecyl phthalate and Diisononyl phthalate; and 3,000 µg/g (3,000 ppm) of Butylbenzyl phthalate, Di-n-butyl phthalate, Diethyl phthalate, Dimethyl phthalate, Di-n-octyl phthalate, and bis(2-Ethylhexyl)phthalate.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixMedium Density Polyethylene
  • Element/IonButylbenzyl phthalate, Di-n-butyl phthalate, Diethyl phthalate, Diisodecyl phthalate, Diisononyl phthalate, Dimethyl phthalate, Di-n-octyl phthalate, bis(2-Ethylhexyl)phthalate
  • FormLiquid
  • Weight (g)5
  • DOT InformationNot Regulated
  • Storage SymbolAmbient
  • Hazard SymbolHealth Hazard
  • UN NumberNot Regulated
  • CAS Number85-68-7, 84-74-2, 84-66-2, 26761-40-0, 28553-12-0, 131-11-3, 117-84-0, 117-81-7
  • Shelf Life (months)36
  • DescriptionPhthalates Polyethylene Standard in Medium Density Polyethylene; 5 g
Documents & Certificates
Reviews
Q&A