Spex CRDL-2A ICP Contract Required Detection Limit (CRDL) Standard 2A; 125 mL

Representative image only.

Spex CRDL-2A ICP Contract Required Detection Limit (CRDL) Standard 2A; 125 mL

Spex | Item: 0456903

Multi-element standards for ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • CLP ISM 01.2 & ILM 05.3 Standards for ICP
  • Multi-element CRDL standard 2A for ICP in 10% HNO3 matrix
  • 125 mL bottle

More About this Item

Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentration levels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:
• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

For ILM 05.3, a standard must be run at the Contract Required Detection Limits (CRDL). To verify linearity near the CRQL, this standard is analyzed at the beginning of the analysis run, after the ICV/ICB and before the ICSA and ICSAB. In addition, this standard must be run at a frequency of not less than 20 analytical samples and at the end of the analysis run, followed by the ICSA/ISCAB. The sequence order is CCV, CCB, CRI, ICSA, ICSAB, CCV, and CCB. For ISM 01.2, at least one of your calibration standards must be at the Contract Required Detection Limit (CRDL). This standard, when diluted, can be used as a calibration standard to fulfill this requirement.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 6 elements.

• ICP Contract Required Detection Limit Standard 2A is a 125 mL bottle which contains 5,000 µg/mL (5,000 ppm) Calcium, Magnesium, Potassium, Sodium and 200 µg/mL (200 ppm) Aluminum, Barium.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix10% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Nitric acid solution; Class 8, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN2031
  • Shelf Life (months)12
  • DescriptionICP Contract Required Detection Limit (CRDL) Standard 2A in 10% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A