Spex CRDL-1 ICP Contract Required Detection Limit (CRDL) Standard 1; 125 mL

Representative image only.

Spex CRDL-1 ICP Contract Required Detection Limit (CRDL) Standard 1; 125 mL

Spex | Item: 0456905

Multi-component CRDL standards for ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • CLP ILM 02.0 & 05.2 Standards for ICP
  • CRDL standard 1 in 5% HNO3/tr. Tartaric Acid/tr. HF matrix
  • 125 mL bottle

More About this Item

Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by Spex CertiPrep for convenience of use and stability.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentration levels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:

• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

CLP ILM 02.0 & 05.2 Standards for ICP
A standard must be run at two times the Contract Required Detection Limits (CRDL), or at two times the Instrument Detection Limits (IDL), whichever is greater. This standardization is performed at the start and the end of each sample analysis or at least twice in each eight hour shift. All elements to be analyzed must be run except Al, Ba, Ca, Fe, K, Mg, and Na. ICP Contract Required Detection Limit (CRDL) Standard 1 (04569-05) contain all of the required elements on the TAL, in their appropriate concentration ratios. ICP Contract Required Detection Limit (CRDL) Standard 1 should be diluted by a factor of 1,000 prior to use in the “two times CRDL” run for ICP-AES analysis. For analysis by atomic absorption, ICP Contract Required Detection Limit (CRDL) Standard 1 should be diluted by a factor of 2,000 prior to use in the “one time CRDL” run.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 15 elements.

• Contract Required Detection Limit standard 1 is a 125 mL bottle of 120 µg/mL (120 ppm) Antimony; 100 µg/mL (100 ppm) Cobalt, Vanadium; 80 µg/mL (80 ppm) Nickel; 50 µg/mL (50 ppm) Copper; 40 µg/mL (40 ppm) Zinc; 30 µg/mL (30 ppm) Manganese; 20 µg/mL (20 ppm) Arsenic, Chromium, Silver, Thallium; 10 µg/mL (10 ppm) Beryllium, Cadmium, Selenium; and 6 µg/mL (6 ppm) Lead in 5% HNO3/tr. Tartaric Acid/tr. HF.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionICP Contract Required Detection Limit (CRDL) Standard 1 in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL
Documents & Certificates
Reviews
Q&A