Spex CLY2-1AM Claritas PPT® Grade Yttrium, 10 µg/mL for ICP-MS in 2% HNO3; 30mL

Representative image only.

Spex CLY2-1AM Claritas PPT® Grade Yttrium, 10 µg/mL for ICP-MS in 2% HNO3; 30mL

Spex | Item: 0450299

Single-element standards for ICP-MS analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with an acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Certified to DQS to ISO 9001:2015
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • 10 ppm yttrium in 2% HNO3 matrix
  • 30 mL bottle

More About this Item

Claritas PPT® grade certified reference materials (CRMs) are designed for inductively coupled plasma (ICP) and ICP-mass spectrometry (MS) analysis. They are made using ultra high purity acids, the highest grade starting materials and high purity water in order to minimize contaminants.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • Purity GradeClaritas PPT® Grade
  • Matrix2% HNO3
  • Element/IonYttrium (Yb)
  • FormLiquid
  • Volume (mL)30
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionClaritas PPT® Grade Yttrium, 10 µg/mL for ICP-MS in 2% HNO3; 30 mL
Documents & Certificates
Reviews
Q&A