Spex CLRH1-1AM Claritas PPT® Grade Rhodium, 10 µg/mL for ICP-MS in 2% HCl; 30 mL

Representative image only.

Spex CLRH1-1AM Claritas PPT® Grade Rhodium, 10 µg/mL for ICP-MS in 2% HCl; 30 mL

Spex | Item: 0450287

Single-element standards for ICP-MS analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • 10 ppm rhodium in 2% HCl matrix
  • 30 mL bottle

More About this Item

Claritas PPT® grade certified reference materials (CRMs) are designed for inductively coupled plasma (ICP) and ICP-mass spectrometry (MS) analysis. Custom standards can be manufactured upon request.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • Purity GradeClaritas PPT® Grade
  • Matrix2% HCl
  • Element/IonRhodium (Rh)
  • FormLiquid
  • Volume (mL)30
  • DOT InformationHazardous; Hydrochloric acid; Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberUN1789
  • Shelf Life (months)12
  • DescriptionClaritas PPT® Grade Rhodium, 10 µg/mL for ICP-MS in 2% HCl; 30 mL
Documents & Certificates
Reviews
Q&A