Spex CLPV-SH-PK Surrogate Standard for EPA Method 8260 & CLP Series Method, 2,500 µg/mL; 1 mL; 4/pk

Representative image only.

Spex CLPV-SH-PK Surrogate Standard for EPA Method 8260 & CLP Series Method, 2,500 µg/mL; 1 mL; 4/pk

Spex | Item: 0500902

Analytical standards for monitoring VOCs by GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8260 and CLP Series Methods
  • 2,500 ppm surrogate standard in methanol-P&T matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixEthanol
  • ChemistryOrganics;Standards
  • Element/IonDi-n-heptyltin dichloride
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Ethanol; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Health Hazard
  • CAS Number74340-12-8
  • UN NumberUN1170
  • Shelf Life (months)12
  • DescriptionDi-n-heptyltin Dichloride Organic Standard, 1,000 µg/mL in Ethanol; 1 mL
Documents & Certificates
Reviews
Q&A