Spex CLPV-D90H Volatiles Mix D for CLP Series Methods, 2,000 µg/mL; 1 mL

Representative image only.

Spex CLPV-D90H Volatiles Mix D for CLP Series Methods, 2,000 µg/mL; 1 mL

Spex | Item: 0455679

Analytical standards for monitoring VOCs by GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 2,000 ppm volatiles mix D in methanol-P&T matrix
  • For CLP Series Methods
  • 1 mL ampule

More About this Item

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • Element/Ion2-Butanone; 2-Hexanone; 4-Methyl-2-pentanone; Acetone; Carbon disulfide; cis-1,2-Dichloroethene; p-Xylene; Styrene
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number78-93-3, 591-78-6, 108-10-1, 67-64-1, 75-15-0, 156-59-2, 106-42-3, 100-42-5
  • UN NumberUN1230
  • Shelf Life (months)6
  • DescriptionVolatiles Mix D for CLP Series Methods, 2,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A