Spex CLPS-SBH5-PK Base/Neutral Surrogate Standard, 5,000 µg/mL; 5 mL; 4/Pk

Representative image only.

Spex CLPS-SBH5-PK Base/Neutral Surrogate Standard, 5,000 µg/mL; 5 mL; 4/Pk

Spex | Item: 0500937

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 625 and CLP Series Methods
  • 5,000 ppm base/neutral surrogate standards in methylene chloride:acetone:benzene matrix
  • 5 mL bottle

More About this Item

US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 3 components.

• Base/Neutral Surrogate Standard is a 5 mL bottle which contains 5,000 µg/mL (5,000 ppm) of 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryOrganics;Standards
  • Element/Ion2,3,5-Trimethylnaphthalene
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number2245-38-7
  • UN NumberUN1230
  • Shelf Life (months)36
  • Description2,3,5-Trimethylnaphthalene Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A