Spex CLPS-SB Base/Neutral Surrogate Standard for EPA & CLP, 1,000 µg/mL; 1 mL

Representative image only.

Spex CLPS-SB Base/Neutral Surrogate Standard for EPA & CLP, 1,000 µg/mL; 1 mL

Spex | Item: 0455790

Analytical standard for monitoring municipal water, industrial wastewater, and environmental and waste samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 625, 8270 and CLP Series Methods
  • 1,000 ppm base/neutral surrogate standards in methylene chloride:acetone matrix
  • 1 mL ampule

More About this Item

US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethylene Chloride:Acetone
  • Element/Ion2-Fluorobiphenyl, Nitrobenzene-d5, p-Terphenyl-d14
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Flammable liquids, n.o.s (Acetone, Dichloromethane); Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • CAS Number321-60-8, 4165-60-0, 1718-51-0
  • UN NumberUN1993
  • Shelf Life (months)36
  • DescriptionBase/Neutral Surrogate Standard for US EPA Methods 625, 8270 and CLP Series Methods, 1,000 µg/mL in Methylene Chloride:Acetone; 1 mL
Documents & Certificates
Reviews
Q&A