Spex CLPS-A-PK Phenolics Mix, 2,000 µg/mL in Methylene Chloride; 1 mL; 4/pk

Representative image only.

Spex CLPS-A-PK Phenolics Mix, 2,000 µg/mL in Methylene Chloride; 1 mL; 4/pk

Spex | Item: 0500928

Analytical standard for municipal water, industrial water, and waste samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 604, 625, 8270 and CLP Series Methods
  • 2,000 ppm phenolic mix in methylene chloride matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 604 is an analytical method for the monitoring of phenols in municipal and industrial wastewater by GC/FID.

US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 11 components.

• Phenolic Mix for US EPA Methods 604, 625, 8270 and CLP Series Methods is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of 2,4,6-Trichlorophenol, 2,4-Dichlorophenol, 2,4-Dimethylphenol, 2,4-Dinitrophenol, 2-Chlorophenol, 2-Nitrophenol, 4,6-Dinitro-2-methylphenol, 4-Chloro-3-methylphenol, 4-Nitrophenol, Pentachlorophenol, and Phenol.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryOrganics;Standards
  • Element/Ionn-Octane
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number111-65-9
  • UN NumberUN1230
  • Shelf Life (months)36
  • Descriptionn-Octane Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A