Spex CLP90-SURR Alternate Combination Surrogates for CLP SOW 3-90; 1 mL

Representative image only.

Spex CLP90-SURR Alternate Combination Surrogates for CLP SOW 3-90; 1 mL

Spex | Item: 0455771

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For CLP Series Methods
  • Organic CLP standard in methylene chloride:acetone matrix
  • 1 mL ampule

More About this Item

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 8 components.

• Alternate Combination Surrogates for CLP SOW 3-90 is a 1 mL ampule which contains 1,500 µg/mL (1,500 ppm) of 2-Chlorophenol-3,4,5,6-d4, 2-Fluorophenol, Phenol-d6, and 2,4,6-Tribromophenol; and 1,000 µg/mL (1,000 ppm) of 1,2-Dichlorobenzene-d4, 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixMethylene Chloride:Acetone
  • Element/Ion2-Chlorophenol-3,4,5,6-d4, 1,2-Dichlorobenzene-d4, 2-Fluorobiphenyl, 2-Fluorophenol, Nitrobenzene-d5, Phenol-d6, p-Terphenyl-d14, 2,4,6-Tribromophenol
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Flammable liquids, n.o.s (Acetone, Dichloromethane); Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1993
  • CAS Number93951-73-6, 2199-69-1, 321-60-8, 367-12-4, 4165-60-0, 13127-88-3, 1718-51-0, 118-79-6
  • Shelf Life (months)24
  • DescriptionAlternate Combination Surrogates for CLP SOW 3-90 in Methylene Chloride:Acetone; 1 mL
Documents & Certificates
Reviews
Q&A