Spex CLP90-SB5 Base/Neutral Surrogate Standard, 1,000 µg/mL; 5 mL

Representative image only.

Spex CLP90-SB5 Base/Neutral Surrogate Standard, 1,000 µg/mL; 5 mL

Spex | Item: 0455769

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For CLP Series Methods
  • 1,000 ppm base/neutral surrogate standards in methylene chloride:acetone matrix
  • 5 mL bottle

More About this Item

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 4 components.

• Base/Neutral Surrogate Standard is a 5 mL bottle which contains 1,000 µg/mL (1,000 ppm) of 1,2-Dichlorobenzene-d4, 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethylene Chloride:Acetone
  • Element/Ion1,2-Dichlorobenzene-d4, 2-Fluorobiphenyl, Nitrobenzene-d5, p-Terphenyl-d14
  • FormLiquid
  • Volume (mL)5
  • DOT InformationHazardous; Flammable liquids, n.o.s (Acetone, Dichloromethane); Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1993
  • CAS Number2199-69-1, 321-60-8, 4165-60-0, 1718-51-0
  • Shelf Life (months)36
  • DescriptionBase/Neutral Surrogate Standards, 1,000 µg/mL in Methylene Chloride:Acetone; 5 mL
Documents & Certificates
Reviews
Q&A