Spex CLMS-5 Claritas PPT® Grade ICP-MS Multi-Element Solution 5, 10 µg/mL; 125 mL

Representative image only.

Spex CLMS-5 Claritas PPT® Grade ICP-MS Multi-Element Solution 5, 10 µg/mL; 125 mL

Spex | Item: 0456887

Multi-element standards for ICP-MS analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Designed to contain virtually every element in the mass spectrum for concentration verification checks
  • 10 ppm multi-element solution 5 in 2% HNO3 matrix
  • 125 mL bottle

More About this Item

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Many of our Claritas PPT® grade multi-element solutions are designed for use with US EPA Methods.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • Purity GradeClaritas PPT® Grade
  • Matrix2% HNO3
  • Element/IonBeryllium, Bismuth, Cerium, Cobalt, Indium, Lead, Magnesium, Nickel, Uranium
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionClaritas PPT® Grade ICP-MS Multi-Element Solution 5, 10 µg/mL in 2% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A