Spex CLHG6-1BY Claritas PPT® Grade Mercury, 1 µg/mL in 0.7% HNO3/0.4% HCl; 125 mL

Representative image only.

Spex CLHG6-1BY Claritas PPT® Grade Mercury, 1 µg/mL in 0.7% HNO3/0.4% HCl; 125 mL

Spex | Item: 0450223

Single-element standards for ICP-MS analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • 1 ppm Mercury in 0.7% HNO3/0.4% HCl matrix

More About this Item

Claritas PPT® grade certified reference materials (CRMs) are designed for inductively coupled plasma (ICP) and ICP-mass spectrometry (MS) analysis. Custom standards can be manufactured upon request.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1 µg/mL (1 ppm)
  • Matrix0.7% HNO3/0.4% HCl
  • Element/IonMercury (Hg)
  • FormLiquid
  • Volume (mL)125
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • UN NumberUN3264
  • Shelf Life (days)180
  • Shelf Life (months)6
  • DescriptionClaritas PPT® Grade Mercury, 1 µg/mL for ICP-MS in 0.7% HNO3/0.4% HCl, 125 mL
Documents & Certificates
Reviews
Q&A