Spex CLBLK-HNO3 Claritas PPT® Grade Nitric Acid Blank in 2% HNO3; 125 mL

Representative image only.

Spex CLBLK-HNO3 Claritas PPT® Grade Nitric Acid Blank in 2% HNO3; 125 mL

Spex | Item: 0456755

Analytical quality matrix blanks ensure highly reliable results

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Run a blank to establish your base line
  • May be used to dilute multi-element standards
  • Nitric acid blank for ICP-MS in 2% HNO3 matrix
  • 125 mL bottle

More About this Item

Do not use any acid or water as a diluent if you are not certain of its purity.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeClaritas PPT® Grade
  • Matrix2% HNO3
  • Element/IonNitric Acid Blank
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionClaritas PPT® Grade Nitric Acid Blank in 2% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A