Spex CLBLK-H2O Claritas PPT® Grade DI Water Blank for ICP-MS in H2O; 125 mL

Representative image only.

Spex CLBLK-H2O Claritas PPT® Grade DI Water Blank for ICP-MS in H2O; 125 mL

Spex | Item: 0456757

Analytical quality matrix blanks ensure highly reliable results

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Run a blank to establish your base line
  • May be used to dilute multi-element standards
  • DI water blank for in H2O matrix
  • 125 mL bottle

More About this Item

Do not use any acid or water as a diluent if you are not certain of its purity.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeClaritas PPT® Grade
  • MatrixH2O
  • Element/IonDI Water Blank
  • FormLiquid
  • Volume (mL)125
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • Polarity Index9
  • Solubility in Water (% wt/wt)100
  • Specific Gravity1
  • UV Cutoff (nm)200
  • DescriptionClaritas PPT® Grade DI Water Blank for ICP-MS in H2O; 125 mL
Documents & Certificates
Reviews
Q&A