Spex CLBA2-2M Claritas PPT® Grade Barium, 1,000 µg/mL in 2% HNO3; 30 mL

Representative image only.

Spex CLBA2-2M Claritas PPT® Grade Barium, 1,000 µg/mL in 2% HNO3; 30 mL

Spex | Item: 0450265

Single-element standards for ICP-MS analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with an acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Certified to DQS to ISO 9001:2015
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • 1,000 ppm barium in 2% HNO3 matrix
  • 30 mL bottle

More About this Item

Claritas PPT® grade certified reference materials (CRMs) are designed for inductively coupled plasma (ICP) and ICP-mass spectrometry (MS) analysis. They are made using ultra high purity acids, the highest grade starting materials and high purity water in order to minimize contaminants.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • Purity GradeClaritas PPT® Grade
  • Matrix2% HNO3
  • Element/IonBarium (Ba)
  • FormLiquid
  • Volume (mL)30
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionClaritas PPT® Grade Barium, 1,000 µg/mL for ICP-MS in 2% HNO3; 30 mL
Documents & Certificates
Reviews
Q&A