Spex CL-MEM-SET Claritas PPT® Grade ICP-MS Memory Test Solution Set; 125 mL

Representative image only.

Spex CL-MEM-SET Claritas PPT® Grade ICP-MS Memory Test Solution Set; 125 mL

Spex | Item: 0456892

Multi-element standard for ICP-MS analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Memory test solution set for ICP-MS contains 2 solutions
  • Two 125 mL bottles

More About this Item

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation.

Use to identify or confirm the maximum concentration of an analyte that does not cause a memory effect greater than the contract required detection limit (CRDL). The test solutions are not analyzed directly; equal volumes of the two are mixed and then introduced into the instrument for a normal sample exposure time. A blank is then run to confirm that all analyte memory effects are below the CRDL.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This memory test solution set contains 2 solutions.

• Solution 1 is a 125 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Aluminum, Calcium, Iron, Magnesium, Potassium, Sodium and 20 µg/mL (20 ppm) of Arsenic, Barium, Beryllium, Cadmium, Chromium, Cobalt, Copper, Lead, Manganese, Nickel, Selenium, Silver, Thallium, Vanadium, Zinc in 5% HNO3. This is available individually as part number 04568-90.

• Solution 2 is a 125 mL bottle which contains 7,200 µg/mL (7,200 ppm) of Chloride; 2,000 µg/mL (2,000 ppm) of Carbon; 1,000 µg/mL (1,000 ppm) of Phosphorus, Sulfur; and 20 µg/mL (20 ppm) of Antimony, Molybdenum, Titanium in H2O/tr. HF. This is available individually as part number 04568-91.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Purity GradeClaritas PPT® Grade
  • MatrixMultiple
  • Element/IonTwo bottle set
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Grup III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionSpex CertiPrep Claritas PPT® Grade ICP-MS Memory Test Solution Set; 125 mL
Reviews
Q&A