Spex CL-MEM-1 Claritas PPT® Grade ICP-MS Memory Test 1 in 5% HNO3; 125 mL

Representative image only.

Spex CL-MEM-1 Claritas PPT® Grade ICP-MS Memory Test 1 in 5% HNO3; 125 mL

Spex | Item: 0456890

Multi-element standards for ICP-MS analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Memory test 1 in 5% HNO3 matrix
  • 125 mL bottle

More About this Item

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation.

Use to identify or confirm the maximum concentration of an analyte that does not cause a memory effect greater than the contract required detection limit (CRDL). The memory test solutions are not analyzed directly; equal volumes of the two (Memory Test 1 and 2) are mixed and then introduced into the instrument for a normal sample exposure time. A blank is then run to confirm that all analyte memory effects are below the CRDL.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 21 elements.

• Memory test 1 standard is a 125 mL bottle of 1,000 µg/mL (1,000 ppm) Aluminum, Calcium, Iron, Magnesium, Potassium, Sodium and 20 µg/mL (20 ppm) Arsenic, Barium, Beryllium, Cadmium, Chromium, Cobalt, Copper, Lead, Manganese, Nickel, Selenium, Silver, Thallium, Vanadium, Zinc in 5% HNO3.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeClaritas PPT® Grade
  • ConcentrationMultiple
  • Matrix5% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionClaritas PPT® Grade ICP-MS Memory Test 1 in 5% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A