Spex CL-INT-A3 ICP-MS Interferents Check Solution A3 (for SOW-846); 125 mL

Representative image only.

Spex CL-INT-A3 ICP-MS Interferents Check Solution A3 (for SOW-846); 125 mL

Spex | Item: 0456924

Multi-element standards for ICP-MS instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Multi-element interferents check solution A3 in 5% HNO3/tr. HF matrix
  • 125 mL bottle

More About this Item

Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by SPEX CertiPrep for convenience of use and stability.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentrationlevels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:

• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

We also provide a solution of alternate interferents and alternate analysis. Alternate interferents A (INT-A2) and alternate analytes B (INT-B2) may be prepared in combination with the INT-A1 and INT-B3 solutions mentioned, or any combination involving the four solutions, depending on the analytes and interferents of interest to you. We provides ICP-MS interferents and interferent check solutions for SW-845.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 12 elements.

• ICP-MS Interferents Check Solution A3 standard (for SOW-846) is a 125 mL bottle which contains 20,000 µg/mL (20,000 ppm) Chloride; 3,000 µg/mL (3,000 ppm) Calcium; 2,500 µg/mL (2,500 ppm) Iron, Sodium; 2,000 µg/mL (2,000 ppm) Carbon; 1,000 µg/mL (1,000 ppm) Aluminum, Magnesium, Phosphorus, Potassium, Sulfur; and 20 µg/mL (20 ppm) Molybdenum, Titanium.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionICP-MS Interferents Check Solution A3 (for SOW-846) in 5% HNO3/tr. HF; 125 mL
Documents & Certificates
Reviews
Q&A