Spex CL-ICV-2 Claritas PPT® Grade ICP-MS Initial Calibration Verification Standard 2, 10 µg/mL (10 ppm) in 2% HNO3/tr. HF, 125 mL

Representative image only.

Spex CL-ICV-2 Claritas PPT® Grade ICP-MS Initial Calibration Verification Standard 2, 10 µg/mL (10 ppm) in 2% HNO3/tr. HF, 125 mL

Spex | Item: 0456866

Multi-element standards for ICP and ICP-MS instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Tune and calibrate ICP-MS instrumentation prior to analysis
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • In 2% HNO3/tr. HF

More About this Item

Spex CertiPrep® Claritas PPT® grade standards are a class of inorganic Certified Reference standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.

Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS. Spex CertiPrep®Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.

Dilute to the concentration appropriate for the instrument with equal parts of Claritas PPT® Nitric Acid Blank and Water Blank. For preparation every two weeks, or as needed.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • Purity GradeClaritas PPT® Grade
  • Matrix2% HNO3/tr. HF
  • ClassICP-MS
  • Element/IonTin, Titanium
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionICP-MS Initial Calibration Verification Standard 2, 10 µg/mL (10 ppm) in 2% HNO3/tr. HF, 125 mL
Documents & Certificates
Reviews
Q&A