Spex CAN-TERP-HA High Abundance Terpene Mix, 10 Component, 5,000 µg/mL in Methanol, 1mL

Representative image only.

Spex CAN-TERP-HA High Abundance Terpene Mix, 10 Component, 5,000 µg/mL in Methanol, 1mL

Spex | Item: 4107157

High concentration analytical standard for terpene analysis contains the most abundant terpenes found in cannabis products

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 5,000 ppm high abundance 10 component terpene mix in methanol matrix
  • 1 mL ampule

More About this Item

This Spex CertiPrep® high abundance terpene mix is a high concentration analytical standard for terpene analysis that contains the most abundant terpenes found in cannabis products. Terpenes are the common term for a large group of compounds that contribute to the flavor and smell of botanicals. The diversity of terpenes and terpenoids is recognized by the range of scents and flavors they produce. The terpene abundances can differ greatly between the strains of cannabis, from major terpenes, such as myrcene, often found in percent levels, to the minor terpenes, such as geraniol, mostly found in ppm levels or smaller. Terpene analysis plays a critical role in the development and QC of cannabis products as it provides a quantitative measure of the product’s flavor and scent profile. Terpene testing and profiling can be difficult since the range of abundance across the profile can vary drastically. A CRM (Certified Reference Material) can provide a basis for more accurate testing results by taking into consideration the natural differences in concentration amongst the various compounds in a terpene profile.

Spex CertiPrep® offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all the common contaminants such as pesticide residues, residual solvents, and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration5,000 µg/mL (5,000 ppm)
  • MatrixMethanol
  • Element/IonTerpene Mix
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard, Skin Sensitivity
  • UN NumberUN1230
  • Shelf Life (months)16
  • DescriptionHigh Abundance Terpene Mix, 10 Component, 5,000 µg/mL in Methanol, 1mL
Reviews
Q&A